siRNA Details
Gene Symbol: Aga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031641.1 | NP_001026811.1 | 3 | 361 | ||
XM_006253111.2 | XP_006253173.1 | 4 | 486 | |||
GenBank mRNA | BC098718.1 | - | 3 | 361 | 7366394 |
