siRNA Details
Gene Symbol: Mtrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039003.1 | NP_001034092.1 | 4 | 307 | ||
XM_006227813.2 | XP_006227875.1 | 6 | 936 | |||
XM_006227814.1 | XP_006227876.1 | 4 | 600 | |||
XM_008758680.1 | XP_008756902.1 | 3 | 1010 | |||
GenBank mRNA | BC100107.1 | - | 4 | 307 | 7379770 |
