siRNA Details
Gene Symbol: Slc35d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106098.2 | NP_001099568.2 | 9 | 724 | ||
XM_006253493.2 | XP_006253555.1 | 9 | 1708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
