siRNA Details
Gene Symbol: Agtpbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106100.1 | NP_001099570.1 | 21 | 2922 | ||
XM_008771435.1 | XP_008769657.1 | 21 | 4306 | |||
XM_008771436.1 | XP_008769658.1 | 21 | 4243 | |||
XM_008771437.1 | XP_008769659.1 | 19 | 2864 | |||
XM_008771438.1 | XP_008769660.1 | 12 | 1922 |
