siRNA Details
Gene Symbol: Uimc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013884.1 | NP_001013906.1 | 9 | 1515 | ||
XM_006253613.2 | XP_006253675.1 | 9 | 1919 | |||
XM_006253616.2 | XP_006253678.1 | 10 | 1558 | |||
GenBank mRNA | BC087149.1 | - | 9 | 1515 | 7131093 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
