siRNA Details
Gene Symbol: Snrnp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106107.1 | NP_001099577.1 | 8 | 892 | ||
XM_006253787.2 | XP_006253849.1 | 8 | 914 | |||
XM_006253788.2 | XP_006253850.1 | 8 | 914 | |||
GenBank mRNA | BC088167.1 | - | 8 | 883 |
