siRNA Details
Gene Symbol: Prpf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011923.1 | NP_001011923.1 | 2 | 983 | ||
GenBank mRNA | BC085927.1 | - | 2 | 983 | 7110150 |

More Information
Other Species Targeted

Gene Ontology Categories:
