siRNA Details
Gene Symbol: Nqo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004214.1 | NP_001004214.1 | 6 | 535 | ||
XM_006253867.2 | XP_006253929.1 | 6 | 786 | |||
XM_006253868.2 | XP_006253930.1 | 5 | 544 | |||
GenBank mRNA | BC079157.1 | - | 6 | 535 | 7128990 |
