siRNA Details
Gene Symbol: Gmds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039606.1 | NP_001034695.1 | 2, 3 | 173 | ||
GenBank mRNA | BC104708.1 | - | 2, 3 | 173 | 7317156 |

More Information
Other Species Targeted

Gene Ontology Categories:
