siRNA Details
Gene Symbol: Aldh5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022851.1 | NP_074042.1 | 7, 8 | 1151 | ||
GenBank mRNA | L34821.1 | - | 7, 8 | 1030 |

More Information
Other Species Targeted

Gene Ontology Categories:
