siRNA Details
Gene Symbol: Gmnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106112.1 | NP_001099582.1 | 5, 6 | 694 | ||
XM_006253927.2 | XP_006253989.1 | 5, 6 | 479 | |||
XM_006253928.2 | XP_006253990.1 | 5, 6 | 417 | |||
GenBank mRNA | FQ219913.1 | - | 2, 3 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
