siRNA Details
Gene Symbol: Trim27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134974.1 | NP_001128446.1 | 5 | 1116 | ||
GenBank mRNA | BC169072.1 | - | 5 | 1116 | 5599836 |

More Information
Other Species Targeted

Gene Ontology Categories:
