siRNA Details
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181627.2 | NP_853658.1 | 678 | |||
GenBank mRNA | AY291062.1 | - | 678 | |||
AY291063.2 | - | 873 | ||||
AY291064.2 | - | 690 | ||||
AY291065.2 | - | 1039 | ||||
AY919342.1 | - | 495 | ||||
AY919343.1 | - | 657 | ||||
FJ517605.1 | - | 550 | ||||
FJ517606.1 | - | 392 | ||||
FQ213378.1 | - | 695 | ||||
FQ213584.1 | - | 695 | ||||
FQ214037.1 | - | 696 | ||||
FQ214498.1 | - | 706 | ||||
FQ227907.1 | - | 686 | ||||
FQ229190.1 | - | 696 | ||||
FQ232927.1 | - | 695 |
