siRNA Details
Gene Symbol: Ggps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB118238.1 | - | 4 | 647 | ||
AB118239.1 | - | 4 | 647 | |||
AB118240.1 | - | 4 | 647 | |||
AB118241.1 | - | 4 | 647 | |||
AB118242.1 | - | 4 | 647 | |||
AB118243.1 | - | 4 | 647 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
