siRNA Details
Gene Symbol: Ggps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007626.2 | NP_001007627.1 | 4 | 1016 | ||
XM_008771699.1 | XP_008769921.1 | 3 | 941 | |||
XM_008771700.1 | XP_008769922.1 | 3 | 963 | |||
GenBank mRNA | AB118238.1 | - | 4 | 1002 | ||
AB118239.1 | - | 4 | 1002 | |||
AB118240.1 | - | 4 | 1002 | |||
BC090327.1 | - | 4 | 1016 | 7125199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
