siRNA Details
Gene Symbol: Mki67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271366.1 | NP_001258295.1 | 13 | 9076 | ||
XM_006230453.2 | XP_006230515.1 | 12 | 9425 | |||
GenBank mRNA | FQ227329.1 | - | 1 | 42 | ||
FQ230341.1 | - | 1 | 42 |
