siRNA Details
Gene Symbol: Zmynd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203366.2 | NP_976242.2 | 10 | 1011 | ||
NM_203367.2 | NP_976243.2 | 10 | 1011 | |||
NM_203368.2 | NP_976244.2 | 9 | 849 | |||
NM_203369.2 | NP_976245.2 | 10 | 930 | |||
XM_006254150.2 | XP_006254212.1 | 12 | 1241 | |||
XM_006254151.2 | XP_006254213.1 | 12 | 1294 | |||
XM_006254152.2 | XP_006254214.1 | 12 | 1153 | |||
XM_006254154.2 | XP_006254216.1 | 11 | 1196 | |||
XM_006254155.2 | XP_006254217.1 | 11 | 1248 | |||
XM_008771704.1 | XP_008769926.1 | 12 | 1291 | |||
XM_008771705.1 | XP_008769927.1 | 12 | 1292 | |||
XM_008771706.1 | XP_008769928.1 | 10 | 984 | |||
XM_008771707.1 | XP_008769929.1 | 10 | 1086 | |||
XM_008771708.1 | XP_008769930.1 | 11 | 1246 | |||
XM_008771709.1 | XP_008769931.1 | 11 | 1244 | |||
XM_008771710.1 | XP_008769932.1 | 10 | 1084 | |||
XM_008771711.1 | XP_008769933.1 | 10 | 1082 | |||
GenBank mRNA | AB162617.1 | - | 9 | 872 | ||
AB162618.1 | - | 9 | 872 | |||
AB162619.1 | - | 8 | 710 | |||
AB162620.1 | - | 8 | 710 | |||
BC065308.1 | - | 10 | 927 | 5599817 |
