siRNA Details
Gene Symbol: Zmynd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203366.2 | NP_976242.2 | 2 | 263 | ||
NM_203367.2 | NP_976243.2 | 2 | 263 | |||
NM_203368.2 | NP_976244.2 | 2 | 263 | |||
NM_203369.2 | NP_976245.2 | 3 | 344 | |||
XM_006254150.2 | XP_006254212.1 | 3 | 448 | |||
XM_006254151.2 | XP_006254213.1 | 3 | 501 | |||
XM_006254152.2 | XP_006254214.1 | 3 | 360 | |||
XM_006254154.2 | XP_006254216.1 | 3 | 448 | |||
XM_006254155.2 | XP_006254217.1 | 3 | 500 | |||
XM_008771704.1 | XP_008769926.1 | 3 | 501 | |||
XM_008771705.1 | XP_008769927.1 | 3 | 499 | |||
XM_008771707.1 | XP_008769929.1 | 3 | 500 | |||
XM_008771708.1 | XP_008769930.1 | 3 | 498 | |||
XM_008771709.1 | XP_008769931.1 | 3 | 499 | |||
XM_008771710.1 | XP_008769932.1 | 3 | 498 | |||
XM_008771711.1 | XP_008769933.1 | 3 | 499 | |||
GenBank mRNA | AB162617.1 | - | 1 | 124 | ||
AB162618.1 | - | 1 | 124 | |||
AB162619.1 | - | 1 | 124 | |||
AB162620.1 | - | 1 | 124 | |||
BC065308.1 | - | 3 | 341 | 5599817 |
