siRNA Details
Gene Symbol: Rbm17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013058.1 | NP_001013076.1 | 11 | 1207 | ||
XM_006254205.2 | XP_006254267.1 | 11 | 1314 | |||
GenBank mRNA | BC079248.1 | - | 11 | 1207 | 7132972 | |
FQ230702.1 | - | 10 | 1067 |
