siRNA Details
Gene Symbol: Fam188a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106122.1 | NP_001099592.1 | 8, 9 | 917 | ||
GenBank mRNA | FQ214325.1 | - | 5, 6 | 588 | ||
FQ214453.1 | - | 3, 4 | 447 |

More Information
Other Species Targeted

