siRNA Details
Gene Symbol: Fam188a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106122.1 | NP_001099592.1 | 14 | 1351 | ||
GenBank mRNA | FQ214325.1 | - | 11 | 1022 | ||
FQ214453.1 | - | 9 | 881 | |||
FQ230167.1 | - | 8 | 710 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_024185, BC027202, AK009891, AK049869, AK193641, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
