siRNA Details
Gene Symbol: Slc39a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106124.1 | NP_001099594.1 | 9 | 1456 | ||
XM_006254285.2 | XP_006254347.1 | 11 | 2092 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc39a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106124.1 | NP_001099594.1 | 9 | 1456 | ||
XM_006254285.2 | XP_006254347.1 | 11 | 2092 |