siRNA Details
Gene Symbol: Atp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106130.1 | NP_001099600.1 | 13 | 1442 | ||
XM_006254983.2 | XP_006255045.1 | 13 | 1485 | |||
XM_008772101.1 | XP_008770323.1 | 11 | 2123 |
