siRNA Details
Gene Symbol: Atp9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106130.1 | NP_001099600.1 | 7, 8 | 868 | ||
XM_006254983.2 | XP_006255045.1 | 7, 8 | 911 | |||
XM_008772101.1 | XP_008770323.1 | 5, 6 | 1549 |
