siRNA Details
Gene Symbol: Dym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106133.2 | NP_001099603.2 | 17 | 2073 | ||
XM_008772103.1 | XP_008770325.1 | 16 | 2135 | |||
XM_008772104.1 | XP_008770326.1 | 16 | 2135 | |||
GenBank mRNA | BC098944.1 | - | 9 | 1083 | ||
BC168151.1 | - | 16 | 2064 | 7459379 | ||
CO558582.1 | - | 3 | 286 |
