siRNA Details
Gene Symbol: Cxxc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079698.1 | NP_001073166.1 | 15 | 1927 | ||
XM_006254917.2 | XP_006254979.1 | 15 | 2228 | |||
GenBank mRNA | BC087117.1 | - | 13 | 2373 | ||
BC128781.1 | - | 15 | 1927 | 7370019 |
