siRNA Details
Gene Symbol: Stard6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007627.1 | NP_001007628.1 | 5 | 461 | ||
XM_006254921.2 | XP_006254983.1 | 6 | 683 | |||
XM_008772111.1 | XP_008770333.1 | 6 | 531 | |||
XM_008772112.1 | XP_008770334.1 | 5 | 486 | |||
GenBank mRNA | AY555189.1 | - | 6 | 631 | ||
BC078976.1 | - | 5 | 461 | 7111945 |
