siRNA Details
Gene Symbol: Cep76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100514.1 | NP_001093984.1 | 5 | 740 | ||
XM_006254858.2 | XP_006254920.1 | 5 | 1031 | |||
XM_008772118.1 | XP_008770340.1 | 4 | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
