siRNA Details
Gene Symbol: Afap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106142.1 | NP_001099612.1 | 6 | 612 | ||
XM_006254794.2 | XP_006254856.1 | 6 | 647 | |||
GenBank mRNA | FQ211638.1 | - | 6 | 577 | ||
FQ231300.1 | - | 630 |
