siRNA Details
Gene Symbol: RGD1312005 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106144.2 | NP_001099614.2 | 3 | 598 | ||
XM_006254754.2 | XP_006254816.1 | 4 | 706 | |||
XM_008772125.1 | XP_008770347.1 | 3 | 673 | |||
GenBank mRNA | BC166445.1 | - | 3 | 598 | 8363471 | |
FQ212918.1 | - | 3 | 340 |
