siRNA Details
Gene Symbol: Spry4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106150.1 | NP_001099620.1 | 2 | 919 | ||
XM_008772067.1 | XP_008770289.1 | 2 | 1776 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
