siRNA Details
Gene Symbol: Cxxc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007628.2 | NP_001007629.1 | 1 | 626 | ||
GenBank mRNA | BC083622.1 | - | 1 | 626 | 7134100 |
Gene Symbol: LOC681300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257539.2 | XP_006257601.1 | 1 | 542 |
