siRNA Details
Gene Symbol: Spata24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025636.2 | NP_001020807.2 | 2, 3 | 184 | ||
XM_006254545.2 | XP_006254607.1 | 2, 3 | 303 | |||
GenBank mRNA | DQ080016.1 | - | 2, 3 | 184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
