siRNA Details
Gene Symbol: Slc25a46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100515.1 | NP_001093985.1 | 8 | 1039 | ||
GenBank mRNA | BC090000.1 | - | 4 | 490 | ||
BC166810.1 | - | 8 | 1039 | 8376140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
