siRNA Details
Gene Symbol: Slc25a46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100515.1 | NP_001093985.1 | 8 | 1101 | ||
GenBank mRNA | BC090000.1 | - | 4 | 552 | ||
BC166810.1 | - | 8 | 1101 | 8376140 |

More Information
Other Species Targeted

Gene Ontology Categories:
