siRNA Details
Gene Symbol: RGD1562997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254439.2 | XP_006254501.1 | 10, 11 | 2333 | ||
XM_006254440.2 | XP_006254502.1 | 9, 10 | 1568 | |||
XM_008771989.1 | XP_008770211.1 | 10, 11 | 2333 | |||
XM_008771990.1 | XP_008770212.1 | 10, 11 | 2333 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001100449.
Gene Ontology Categories:
