siRNA Details
Gene Symbol: Snrpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106163.1 | NP_001099633.1 | 2 | 158 | ||
XM_008771943.1 | XP_008770165.1 | 2 | 173 | |||
GenBank mRNA | BC167095.1 | - | 2 | 51 | 9091450 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
