siRNA Details
Gene Symbol: Polr2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134789.1 | NP_001128261.1 | 2 | 237 | ||
GenBank mRNA | CB758009.1 | - | 2 | 251 | ||
CF113682.1 | - | 2 | 210 | |||
FQ222889.1 | - | 2 | 235 |
Gene Symbol: LOC100912534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751083.2 | XP_003751131.1 | 2 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
