siRNA Details
Gene Symbol: RGD1565635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254394.2 | XP_006254456.1 | 3 | 504 | ||
XM_006254395.2 | XP_006254457.1 | 4 | 639 | |||
XM_008771959.1 | XP_008770181.1 | 3 | 431 |
