siRNA Details
Gene Symbol: Cmtm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106164.1 | NP_001099634.1 | 3 | 400 | ||
XM_006255049.2 | XP_006255111.1 | 4 | 483 | |||
XM_008772241.1 | XP_008770463.1 | 4 | 487 | |||
XM_008772242.1 | XP_008770464.1 | 4 | 436 | |||
GenBank mRNA | BC169107.1 | - | 4 | 492 | 7130835 |
