siRNA Details
Gene Symbol: Car7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106165.1 | NP_001099635.1 | 5 | 586 | ||
XM_006255051.2 | XP_006255113.1 | 5 | 714 | |||
XM_008772243.1 | XP_008770465.1 | 4 | 452 | |||
GenBank mRNA | BC167038.1 | - | 5 | 555 | 9090629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
