siRNA Details
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003705.2 | NP_001003705.1 | 7 | 1144 | ||
XM_006255067.2 | XP_006255129.1 | 9 | 1291 | |||
GenBank mRNA | AY294328.1 | - | 8 | 1239 | ||
BC086369.1 | - | 7 | 1144 | 7109456 |
