siRNA Details
Gene Symbol: Nudt21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039004.1 | NP_001034093.1 | 2 | 211 | ||
XM_006255132.2 | XP_006255194.1 | 2 | 844 | |||
GenBank mRNA | BC098748.1 | - | 2 | 211 | 7365565 | |
BC168149.1 | - | 2 | 850 | 7457021 | ||
FQ213930.1 | - | 2 | 254 | |||
FQ229498.1 | - | 2 | 252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
