siRNA Details
Gene Symbol: Nudt21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039004.1 | NP_001034093.1 | 4, 5 | 552 | ||
XM_006255132.2 | XP_006255194.1 | 4, 5 | 1185 | |||
GenBank mRNA | BC098748.1 | - | 4, 5 | 552 | 7365565 | |
BC168149.1 | - | 4, 5 | 1191 | 7457021 | ||
FQ213930.1 | - | 4, 5 | 595 | |||
FQ229498.1 | - | 4, 5 | 593 |
