siRNA Details
Gene Symbol: Mcm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106170.1 | NP_001099640.1 | 2 | 333 | ||
XM_008772283.1 | XP_008770505.1 | 3 | 295 | |||
GenBank mRNA | BC166442.1 | - | 2 | 236 | 8362933 |
