siRNA Details
Gene Symbol: Mmaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106174.1 | NP_001099644.1 | 3 | 605 | ||
XM_006255398.2 | XP_006255460.1 | 2 | 287 | |||
XM_006255399.2 | XP_006255461.1 | 2 | 2426 |

More Information
Other Species Targeted

Gene Ontology Categories:
