siRNA Details
Gene Symbol: Ttc29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013890.1 | NP_001013912.1 | 11 | 1481 | ||
GenBank mRNA | BC078968.1 | - | 11 | 1481 | 7111580 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ttc29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013890.1 | NP_001013912.1 | 11 | 1481 | ||
GenBank mRNA | BC078968.1 | - | 11 | 1481 | 7111580 |