siRNA Details
Gene Symbol: Hsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255453.2 | XP_006255515.1 | 2 | 665 | ||
XM_006255454.2 | XP_006255516.1 | 2 | 665 |

More Information
Other Species Targeted

Gene Ontology Categories:
