siRNA Details
Gene Symbol: Kctd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772558.1 | XP_008770780.1 | 13 | 2407 | ||
XM_226403.8 | XP_226403.6 | 13 | 2472 |

More Information
Other Species Targeted

Gene Ontology Categories:
