siRNA Details
Gene Symbol: Tppp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009639.1 | NP_001009639.1 | 2, 3 | 302 | ||
XM_006255464.2 | XP_006255526.1 | 2, 3 | 332 | |||
GenBank mRNA | BC087585.1 | - | 2, 3 | 302 | 7317980 | |
FQ213895.1 | - | 1, 2 | 444 | |||
FQ214516.1 | - | 2, 3 | 316 | |||
FQ214740.1 | - | 2, 3 | 306 | |||
FQ216797.1 | - | 2, 3 | 327 | |||
FQ217207.1 | - | 2, 3 | 303 |
